Learn More
Selleck Chemical LLC 2-Ketoglutaric acid
SDP
Catalog No. 5000003767
Encompass_Preferred
2-Ketoglutaric acid (alpha-ketoglutarate) is a key molecule in the TCA cycle playing a fundamental role in determining the overall rate of this important metabolic process In the TCA cycle AKG is decarboxylated to succinyl-CoA and carbon dioxide by AKG dehydrogenase which functions as a key control point of the TCA cycle 2-Ketoglutaric acid is a reversible inhibitor of tyrosinase
Catalog No. 50-000-03767 Supplier Selleck Chemical LLC Supplier No. S62375G
2-Ketoglutaric acid (alpha-ketoglutarate) is a key molecule in the TCA cycle playing a fundamental role in determining the overall rate of this important metabolic process In the TCA cycle AKG is decarboxylated to succinyl-CoA and carbon dioxide by AKG dehydrogenase which functions as a key control point of the TCA cycle 2-Ketoglutaric acid is a reversible inhibitor of tyrosinase