Learn More
Medchemexpress LLC 2-Ketoglutaric acid-13C | 108395-15-9 | 97.9% | 147.09 | 500 UG
SDP
Catalog No. 5000454218
Encompass_Preferred
2-Ketoglutaric acid-13C is a 13C labeled 2-Ketoglutaric acid. It is an intermediate in the production of ATP or GTP in the Krebs cycle and serves as the primary carbon skeleton for nitrogen-assimilatory reactions. This compound can be utilized as a tracer or as an internal standard for quantitative analysis. Stable heavy isotopes have been incorporated into drug molecules as tracers for quantitation during drug development.
  • 13C labeled compound
  • Intermediate in ATP or GTP production in Krebs cycle
  • Primary carbon skeleton for nitrogen-assimilatory reactions
  • Reversible inhibitor of tyrosinase (IC50 of 15 mM)
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
Catalog No. 50-004-54218 Supplier Medchemexpress LLC Supplier No. HYW013636S3500UG
2-Ketoglutaric acid-13C is a 13C labeled 2-Ketoglutaric acid. It is an intermediate in the production of ATP or GTP in the Krebs cycle and serves as the primary carbon skeleton for nitrogen-assimilatory reactions. This compound can be utilized as a tracer or as an internal standard for quantitative analysis. Stable heavy isotopes have been incorporated into drug molecules as tracers for quantitation during drug development.
  • 13C labeled compound
  • Intermediate in ATP or GTP production in Krebs cycle
  • Primary carbon skeleton for nitrogen-assimilatory reactions
  • Reversible inhibitor of tyrosinase (IC50 of 15 mM)
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)