Learn More
Medchemexpress LLC 2-Ketoglutaric acid-13C5 | 161096-83-9 | 99.1% | 151.06 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW013636S10MG
Encompass_Preferred
2-Ketoglutaric acid-13C5 is a 13C labeled 2-Ketoglutaric acid that functions as an intermediate in the Krebs cycle for ATP or GTP production and as a major carbon skeleton for nitrogen-assimilatory reactions. It is also a reversible inhibitor of tyrosinase (IC50=15 mM).
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-54463