Learn More
Medchemexpress LLC HY-W013636 1g Medchemexpress, 2-Ketoglutaric acid CAS:328-50-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW0136361G
Encompass_Preferred
Medchemexpress, HY-W013636 1g 2-Ketoglutaric acid CAS:328-50-7 2-Ketoglutaric acid is an intermediate compound of the Krebs cycle and is also connected to glutamic acid and glutamine metabolisms through the transamination reactions. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4585