Learn More
Medchemexpress LLC 2-Ketoglutaric acid | 328-50-7 | 99.68% | 146.10 | 1MG
SDP
Catalog No. 5000454206
Encompass_Preferred
2-Ketoglutaric acid (Alpha-Ketoglutaric acid) is an intermediate involved in the production of ATP or GTP within the Krebs cycle. It also serves as the primary carbon skeleton for nitrogen-assimilatory reactions. Additionally, 2-Ketoglutaric acid functions as a reversible inhibitor of tyrosinase, with an IC50 of 15 mM.
  • Reduction of ammonia levels formed in the lung and general ammonia detoxification.
  • Protective role against lipid peroxidation.
  • Neuroprotective effect against cyanide poisoning.
  • Acts as a precursor for the synthesis of amino acids and nucleotides.
Catalog No. 50-004-54206 Supplier Medchemexpress LLC Supplier No. HYW013636S11MG
2-Ketoglutaric acid (Alpha-Ketoglutaric acid) is an intermediate involved in the production of ATP or GTP within the Krebs cycle. It also serves as the primary carbon skeleton for nitrogen-assimilatory reactions. Additionally, 2-Ketoglutaric acid functions as a reversible inhibitor of tyrosinase, with an IC50 of 15 mM.
  • Reduction of ammonia levels formed in the lung and general ammonia detoxification.
  • Protective role against lipid peroxidation.
  • Neuroprotective effect against cyanide poisoning.
  • Acts as a precursor for the synthesis of amino acids and nucleotides.