Learn More
Medchemexpress LLC 2-Ketoglutaric acid-d6 | 1173021-86-7 | 98.6% | 152.14 | 10 MG
SDP
Catalog No. 5000454603
Encompass_Preferred
2-Ketoglutaric acid-d6 is the deuterium labeled variant of 2-Ketoglutaric acid. It is an intermediate in the Krebs cycle for ATP or GTP production and serves as the primary carbon skeleton for nitrogen-assimilatory reactions. This compound also acts as a reversible inhibitor of tyrosinase (IC50=15 mM).
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated for potential impact on pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-54603 Supplier Medchemexpress LLC Supplier No. HYW013636S210MG
2-Ketoglutaric acid-d6 is the deuterium labeled variant of 2-Ketoglutaric acid. It is an intermediate in the Krebs cycle for ATP or GTP production and serves as the primary carbon skeleton for nitrogen-assimilatory reactions. This compound also acts as a reversible inhibitor of tyrosinase (IC50=15 mM).
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated for potential impact on pharmacokinetic and metabolic profiles of drugs.