Learn More
Alkali Scientific 2 Ketoglutaric Acid Monopotassium Salt, 100 G
SDP
Catalog No. 5000661634
Encompass_Preferred
2-Ketoglutaric Acid Monopotassium Salt (100g) is a versatile reagent used in laboratory experiments requiring a stable and reliable source of this important metabolic intermediate. 2-Ketoglutarate plays a vital role in the Krebs cycle and is involved in amino acid metabolism, making this reagent essential for metabolic studies and research into cellular energy production. This monopotassium salt form is widely used in biochemical assays, enzyme catalysis, and as a precursor for the synthesis of various compounds. This 100g formulation is suitable for large-scale experiments and is widely used in analytical, pharmacological, and biochemical research labs.
Catalog No. 50-006-61634 Supplier Alkali Scientific Supplier No. TOG002CP
2-Ketoglutaric Acid Monopotassium Salt (100g) is a versatile reagent used in laboratory experiments requiring a stable and reliable source of this important metabolic intermediate. 2-Ketoglutarate plays a vital role in the Krebs cycle and is involved in amino acid metabolism, making this reagent essential for metabolic studies and research into cellular energy production. This monopotassium salt form is widely used in biochemical assays, enzyme catalysis, and as a precursor for the synthesis of various compounds. This 100g formulation is suitable for large-scale experiments and is widely used in analytical, pharmacological, and biochemical research labs.