Learn More
Alkali Scientific 2 Ketoglutaric Acid Monopotassium Salt, 25 G
SDP
Catalog No. 5000661277
Encompass_Preferred
2-Ketoglutaric Acid Monopotassium Salt (25g) is an important biochemical reagent used in laboratory studies related to cellular metabolism and enzymology. As a key intermediate in the Krebs cycle, 2-ketoglutarate is crucial for energy production and amino acid biosynthesis in living organisms. This reagent is commonly used in metabolic research, enzymatic reactions, and the synthesis of various compounds. The 25g bottle provides a convenient quantity for small to medium-scale laboratory experiments, making it ideal for biochemical assays and for use in research focused on cellular respiration, energy metabolism, and the role of ketoglutarate in biochemistry.
Catalog No. 50-006-61277 Supplier Alkali Scientific Supplier No. TOG003CP
2-Ketoglutaric Acid Monopotassium Salt (25g) is an important biochemical reagent used in laboratory studies related to cellular metabolism and enzymology. As a key intermediate in the Krebs cycle, 2-ketoglutarate is crucial for energy production and amino acid biosynthesis in living organisms. This reagent is commonly used in metabolic research, enzymatic reactions, and the synthesis of various compounds. The 25g bottle provides a convenient quantity for small to medium-scale laboratory experiments, making it ideal for biochemical assays and for use in research focused on cellular respiration, energy metabolism, and the role of ketoglutarate in biochemistry.