Learn More
Alkali Scientific 2 Ketoglutaric Acid Monopotassium Salt, 5 G
SDP
Catalog No. 5000660513
Encompass_Preferred
2-Ketoglutaric Acid Monopotassium Salt (5g) is a highly purified reagent used in laboratory experiments studying metabolic pathways, particularly the Krebs cycle. 2-Ketoglutarate is a critical compound involved in cellular respiration and amino acid metabolism, making this reagent essential in biochemical and metabolic research. The monopotassium salt form is stable and suitable for a variety of applications, including enzyme catalysis, metabolic assays, and organic synthesis. This 5g quantity is perfect for small-scale experiments in laboratories focused on studying energy production, amino acid biosynthesis, and other metabolic processes.
Catalog No. 50-006-60513 Supplier Alkali Scientific Supplier No. TOG004CP
2-Ketoglutaric Acid Monopotassium Salt (5g) is a highly purified reagent used in laboratory experiments studying metabolic pathways, particularly the Krebs cycle. 2-Ketoglutarate is a critical compound involved in cellular respiration and amino acid metabolism, making this reagent essential in biochemical and metabolic research. The monopotassium salt form is stable and suitable for a variety of applications, including enzyme catalysis, metabolic assays, and organic synthesis. This 5g quantity is perfect for small-scale experiments in laboratories focused on studying energy production, amino acid biosynthesis, and other metabolic processes.