Learn More
Medchemexpress LLC 2-Ketoglutaric acid potassium | 997-43-3 | 98.0% | 185.20 | 500 MG
SDP
Catalog No. 5000454521
Encompass_Preferred
2-Ketoglutaric acid potassium, also known as Alpha-Ketoglutaric acid potassium, is an intermediate in the production of ATP or GTP in the Krebs cycle. It is a reversible and orally active inhibitor of tyrosinase, and also acts as a major carbon skeleton for nitrogen-assimilatory reactions.
  • Suppresses abnormal intestinal permeability.
  • Inhibits delocalization of tight junction proteins from intestinal cells.
  • Reduces expression of TNFα in vitro and in vivo.
  • Directly binds to TAK1 and inhibits TRAF6-TAK1 interaction.
  • Alleviates inflammatory bowel disease (IBD) symptoms and gut microbiota dysbiosis.
Catalog No. 50-004-54521 Supplier Medchemexpress LLC Supplier No. HYW013636C500MG
2-Ketoglutaric acid potassium, also known as Alpha-Ketoglutaric acid potassium, is an intermediate in the production of ATP or GTP in the Krebs cycle. It is a reversible and orally active inhibitor of tyrosinase, and also acts as a major carbon skeleton for nitrogen-assimilatory reactions.
  • Suppresses abnormal intestinal permeability.
  • Inhibits delocalization of tight junction proteins from intestinal cells.
  • Reduces expression of TNFα in vitro and in vivo.
  • Directly binds to TAK1 and inhibits TRAF6-TAK1 interaction.
  • Alleviates inflammatory bowel disease (IBD) symptoms and gut microbiota dysbiosis.