Learn More
Medchemexpress LLC 2-Ketoglutaric acid (Sodium) | 22202-68-2 | 98.0% | 168.08 | 1 ML
SDP
Catalog No. 5000454594
Encompass_Preferred
2-Ketoglutaric acid Sodium (Alpha-Ketoglutaric acid Sodium) is an intermediate involved in the production of ATP or GTP within the Krebs cycle. It also functions as the primary carbon skeleton for nitrogen-assimilatory reactions. Additionally, 2-Ketoglutaric acid Sodium is a reversible inhibitor of tyrosinase, with an IC50 of 15 mM.
  • Intermediate in ATP/GTP production in the Krebs cycle.
  • Major carbon skeleton for nitrogen-assimilatory reactions.
  • Reversible inhibitor of tyrosinase (IC50=15 mM).
  • Reduces ammonia levels formed in the lung and aids in general ammonia detoxification.
  • Offers protective effects against lipid peroxidation and neuroprotection against cyanide poisoning.
  • Acts as a precursor for the synthesis of amino acids and nucleotides.
Catalog No. 50-004-54594 Supplier Medchemexpress LLC Supplier No. HYW013636A10MM/1ML
2-Ketoglutaric acid Sodium (Alpha-Ketoglutaric acid Sodium) is an intermediate involved in the production of ATP or GTP within the Krebs cycle. It also functions as the primary carbon skeleton for nitrogen-assimilatory reactions. Additionally, 2-Ketoglutaric acid Sodium is a reversible inhibitor of tyrosinase, with an IC50 of 15 mM.
  • Intermediate in ATP/GTP production in the Krebs cycle.
  • Major carbon skeleton for nitrogen-assimilatory reactions.
  • Reversible inhibitor of tyrosinase (IC50=15 mM).
  • Reduces ammonia levels formed in the lung and aids in general ammonia detoxification.
  • Offers protective effects against lipid peroxidation and neuroprotection against cyanide poisoning.
  • Acts as a precursor for the synthesis of amino acids and nucleotides.