Learn More
Medchemexpress LLC 2-Ketoglutaric acid (standard) | 328-50-7 | 99.5% | 146.10 | 50 MG
SDP
Catalog No. 5000454462
Encompass_Preferred
2-Ketoglutaric acid (Standard) is the analytical standard of 2-Ketoglutaric acid. This product is intended for research and analytical applications. It is an intermediate in the production of ATP or GTP in the Krebs cycle and acts as the major carbon skeleton for nitrogen-assimilation reactions. 2-Ketoglutaric acid is a reversible inhibitor of tyrosinase (IC50=15 mM).
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-004-54462 Supplier Medchemexpress LLC Supplier No. HYW013636R50MG
2-Ketoglutaric acid (Standard) is the analytical standard of 2-Ketoglutaric acid. This product is intended for research and analytical applications. It is an intermediate in the production of ATP or GTP in the Krebs cycle and acts as the major carbon skeleton for nitrogen-assimilation reactions. 2-Ketoglutaric acid is a reversible inhibitor of tyrosinase (IC50=15 mM).
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.