Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ 2 KOH Coated Silica Gel

Catalog No. 111001614 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-1614 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-1614 Supplier MilliporeSigma™ Supelco™ Supplier No. 21318U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Treated silica gels may be primarily employed in the pre-packed multi-layer silica gel columns are available in bulk packages.

Treated silica gels may be primarily employed in the pre-packed multi-layer silica gel columns are available in bulk packages. These are attractive options for customizing the required columns to more efficiently clean very dirty samples, or to prepare shorter columns when samples are relatively clean, e.g. drinking water.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.