Learn More
MSC Miller StretchStop Shock-Absorbing Lanyard, Webbing, Two-Leg, 6 ft., 1 ea.
Catalog No. 5000509818
Encompass
This stretchable two-legged shock-absorbing lanyard is engineered for fall protection, providing strength and comfort. The system utilizes a specially-woven inner core that expands to reduce fall arrest forces, and features corrosion-resistant hardware for reliability in demanding conditions.
  • Includes SofStop® shock absorber with vinyl cover
  • Features two legs for continuous tie-off
  • Equipped with steel hardware tested to fifty-hour ASTM salt spray standards
  • Constructed from durable Type 7 webbing
  • Includes locking snap hook and locking rebar hook
  • Meets ANSI Z359.13-2009, OSHA, and CSA Z259.11-17 standards
  • Maximum working load capacity of 310 lbs.
Catalog No. 50-005-09818 Supplier MSC Supplier No. 58140955
This stretchable two-legged shock-absorbing lanyard is engineered for fall protection, providing strength and comfort. The system utilizes a specially-woven inner core that expands to reduce fall arrest forces, and features corrosion-resistant hardware for reliability in demanding conditions.
  • Includes SofStop® shock absorber with vinyl cover
  • Features two legs for continuous tie-off
  • Equipped with steel hardware tested to fifty-hour ASTM salt spray standards
  • Constructed from durable Type 7 webbing
  • Includes locking snap hook and locking rebar hook
  • Meets ANSI Z359.13-2009, OSHA, and CSA Z259.11-17 standards
  • Maximum working load capacity of 310 lbs.