Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) PTC22010 Coupling Insert, Two-Line, Acetal, 1/16"" ID Tube
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3105004
Encompass
Colder has made it easy to handle multiple tubing lines in your process that require connection and reconnection! These multiline couplings simplify the manual steps in your process for quick transfers and minimize your down time. Twin Tube™ series: With two lines in one fitting, you can connect or disconnect with the push of a button. Each line is capable of independently handling anything from vacuum to 120 psi. The coupling allows rotation of both tubing lines to prevent kinking. The Twin Tube™ series is available with hose barb connections only. Sixtube™ series: With a single connector, control six different lines! An efficient thumblatch design allows quick disconnecting with optional shutoff.
Catalog No. 50-628-280