Learn More
Cayman Chemical 2-LInoleoyl Glycerol 1mg
SDP
Catalog No. 502896251
Encompass_Preferred
A congener of 2-AG in which a linoleoyl group replaces the arachidonoyl group that potentiates the activity of other endocannabinoids, including 2-AG; this “entourage” effect has been attributed to blockade of the breakdown and reuptake pathways that normally function to rapidly reduce endocannabinoid levels following release
Catalog No. 50-289-6251 Supplier Cayman Chemical Supplier No. 622601 MG
A congener of 2-AG in which a linoleoyl group replaces the arachidonoyl group that potentiates the activity of other endocannabinoids, including 2-AG; this “entourage” effect has been attributed to blockade of the breakdown and reuptake pathways that normally function to rapidly reduce endocannabinoid levels following release