Learn More
Medchemexpress LLC 2-Linoleoyl glycerol (2-Monolinolein) | 3443-82-1 | 99.9% | 1 MG
SDP
Catalog No. 5000340940
Encompass_Preferred
2-Linoleoyl glycerol (2-Monolinolein) is a monoacylglycerol that acts as an antagonist and partial agonist at the type 1 cannabinoid CB1 receptor. Its potency can be increased by JZL195, an inhibitor of FAAH and MAGL, and decreased by the CB1 antagonist AM251 and Cannabidiol. As a CB1 antagonist, it attenuates the activity of CB1/CB2 receptor ligands such as cannabinoids (AEA) and 2-arachidonoylglycerol (2-AG).
  • Acts as an antagonist and partial agonist at the type 1 cannabinoid CB1 receptor
  • Potency can be enhanced by JZL195
  • Activity can be inhibited by AM251 and Cannabidiol
  • Attenuates the activity of CB1/CB2 receptor ligands
Catalog No. 50-003-40940 Supplier Medchemexpress LLC Supplier No. HY1303111MG
2-Linoleoyl glycerol (2-Monolinolein) is a monoacylglycerol that acts as an antagonist and partial agonist at the type 1 cannabinoid CB1 receptor. Its potency can be increased by JZL195, an inhibitor of FAAH and MAGL, and decreased by the CB1 antagonist AM251 and Cannabidiol. As a CB1 antagonist, it attenuates the activity of CB1/CB2 receptor ligands such as cannabinoids (AEA) and 2-arachidonoylglycerol (2-AG).
  • Acts as an antagonist and partial agonist at the type 1 cannabinoid CB1 receptor
  • Potency can be enhanced by JZL195
  • Activity can be inhibited by AM251 and Cannabidiol
  • Attenuates the activity of CB1/CB2 receptor ligands