Learn More
Thermo Scientific 2 LITER PLASTIC BOTTLES WITH C
Catalog No. NC9289297 Shop All Thermo Scientific Products
Encompass
Used in displacement chemical regeneration operation for Dionex AMMS™ or CMMS™ MicroMembrane™ Suppressors. Eluent Bottle; 2L; Plastic; Each. ◾Non-twist stopper with separate retaining ring prevents tubing from tangling ◾Graduations marked on containers in 100mL increments ◾Includes custom-molded polyethylene end-line filters with 5μm pores
Catalog No. NC9289297 Supplier Thermo Scientific Supplier No. 044129
Used in displacement chemical regeneration operation for Dionex AMMS™ or CMMS™ MicroMembrane™ Suppressors. Eluent Bottle; 2L; Plastic; Each. ◾Non-twist stopper with separate retaining ring prevents tubing from tangling ◾Graduations marked on containers in 100mL increments ◾Includes custom-molded polyethylene end-line filters with 5μm pores