Learn More
Cole-Parmer Sherwood Scientific 50035006 Fluid Bed Dryer Tub and Assembly, Non-Sealed, Glass, 2-Liter
Catalog No. NC1252990 Shop All Cole-Parmer Products
Encompass_Preferred
Glass tubs are useful for observation of applications and determination of the optimum fluidizing velocity.
Catalog No. NC1252990 Supplier Cole-Parmer Supplier No. 0265852
Glass tubs are useful for observation of applications and determination of the optimum fluidizing velocity.