Learn More
Medchemexpress LLC 2-Maleimido acetic acid | 25021-08-3 | MFCD00457254 | 100.0% | 155.11 g/mol | C6H5NO4 | 1 ML
SDP
Catalog No. 5000301312
Encompass_Preferred
2-Maleimido acetic acid is a carboxylic acid bearing a maleimide functional group used as a bioconjugation reagent and synthetic intermediate. It reacts selectively with thiol groups, enabling site-specific conjugation of proteins, peptides, and polymers, and is offered as solid material or as a ready-to-use solution for convenient use in assays and synthesis.
  • Contains an active maleimide group for selective thiol coupling.
  • Enables site-specific conjugation of cysteine residues in biomolecules.
  • Available as a ready-to-use 10 mM solution in DMSO for rapid use.
  • Also supplied as solid material in multiple milligram pack sizes.
  • High purity suitable for synthesis and analytical applications.
Catalog No. 50-003-01312 Supplier Medchemexpress LLC Supplier No. HYW01783110MM/1ML
2-Maleimido acetic acid is a carboxylic acid bearing a maleimide functional group used as a bioconjugation reagent and synthetic intermediate. It reacts selectively with thiol groups, enabling site-specific conjugation of proteins, peptides, and polymers, and is offered as solid material or as a ready-to-use solution for convenient use in assays and synthesis.
  • Contains an active maleimide group for selective thiol coupling.
  • Enables site-specific conjugation of cysteine residues in biomolecules.
  • Available as a ready-to-use 10 mM solution in DMSO for rapid use.
  • Also supplied as solid material in multiple milligram pack sizes.
  • High purity suitable for synthesis and analytical applications.