Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Maleimidoethyl Mesylate

Catalog No. 413141GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-314-1GF 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 41-314-1GF Supplier Sigma Aldrich Supplier No. 413141GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hetero-bifunctional linker specially for the quaternisation of pyridyl-nitrogen in order to introduce an amine or thiol reactive group. Synthetic building block for the preparation of fluorescent labels

Specifications

CAS 70755-47-4
Melting Point 159°C to 162°C
Molecular Formula C34H72BrN
Linear Formula [CH3(CH2)15]2N(CH3)2(Br)
MDL Number MFCD00191428
Quantity 1 g
Molecular Weight (g/mol) 574.85 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.