Learn More
Cayman Chemical 2-MAPBhydrochlorIde 10mg
SDP
Catalog No. 502967755
Encompass_Preferred
An analytical reference standard categorized as a benzofuran; has been associated with fatal overdoses; intended for research and forensic applications
Catalog No. 50-296-7755 Supplier Cayman Chemical Supplier No. 1714010 MG
An analytical reference standard categorized as a benzofuran; has been associated with fatal overdoses; intended for research and forensic applications