Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Mercapto-1-methylimidazole

Catalog No. 501764273
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-176-4273 25 g
50-176-4274 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-176-4273 Supplier Sigma Aldrich Supplier No. 30150725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Mercapto-1-methylimidazole
TRUSTED_SUSTAINABILITY

Specifications

CAS 60-56-0
Melting Point 144°C to 147°C (lit.)
Molecular Formula C4H6N2S
Linear Formula C4H6N2S
MDL Number MFCD00179321
Quantity 25 g
Synonym Methimazole; 1-Methyl-2-imidazolethiol; 2-Mercapto-1-methylimidazole
Molecular Weight (g/mol) 114.17
Percent Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.