Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercapto-1-methylimidazole, 98%

Catalog No. AC125500250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC125500250 25 g Glass bottle
AC125501000 100 g Glass bottle
AC125505000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC125500250 Supplier Thermo Scientific Chemicals Supplier No. 125500250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

2-Mercapto-1-methylimidazole, CAS # 60-56-0, also known as methimazole, is a thyroid hormone antagonist used to study the treatment of thyroid disorders. | CAS: 60-56-0 | C4H6N2S | 114.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • 2-Mercapto-1-methylimidazole, also known as methimazole, is an antithyroid medication
  • Methimazole works by inhibiting the enzyme thyroid peroxidase (TPO), which is responsible for synthesizing the thyroid hormones triiodothyronine (T3) and thyroxine (T4) from the precursor thyroglobulin

Applications

  • Methimazole has been used in laboratory settings to model olfactory epithelial degeneration
  • May be used to study disruptions in iodide metabolism
  • Methimazole has been used in Pharmacokinetics studies in the laboratory
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-56-0
Molecular Formula C4H6N2S
Molecular Weight (g/mol) 114.17
MDL Number MFCD00179321
InChI Key PMRYVIKBURPHAH-UHFFFAOYSA-N
Synonym methimazole, thiamazole, 2-mercapto-1-methylimidazole, tapazole, mercazolyl, 1-methylimidazole-2-thiol, mercazole, metazolo, methimazol, thiamazol
PubChem CID 1349907
ChEBI CHEBI:50673
IUPAC Name 3-methyl-1H-imidazole-2-thione
SMILES CN1C=CNC1=S

Specifications

Melting Point 140.0°C to 145.0°C
Color Cream to White
Boiling Point 280.0°C
Flash Point >100°C
Loss on Drying 1% (105°C, 2 hrs) max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 24,17
Merck Index 15,6043
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and acetone,sparingly soluble in ether,petr. ether,benzene
Formula Weight 114.16
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Mercapto-1-methylimidazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Contaminated work clothing should not be allowed out of the workplace.
EINECSNumber 200-482-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.