Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercapto-1-methylimidazole 98.0+%, TCI America™
SDP

Catalog No. M086825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M086825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M086825G Supplier TCI America Supplier No. M086825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 60-56-0
Molecular Formula C4H6N2S
Molecular Weight (g/mol) 114.17
MDL Number MFCD00179321
InChI Key PMRYVIKBURPHAH-UHFFFAOYSA-N
Synonym methimazole, thiamazole, 2-mercapto-1-methylimidazole, tapazole, mercazolyl, 1-methylimidazole-2-thiol, mercazole, metazolo, methimazol, thiamazol
PubChem CID 1349907
ChEBI CHEBI:50673
IUPAC Name 1-methyl-2,3-dihydro-1H-imidazole-2-thione
SMILES CN1C=CNC1=S

Specifications

Melting Point 145°C
Color White
Quantity 25 g
Formula Weight 114.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Mercapto-1-methylimidazole
EINECSNumber (9)-1433
RTECSNumber NI8615000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.