Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Mercapto-5-benzimidazolesulfonic acid sodium salt dihydrate

Catalog No. 38570025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-570-025G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-570-025G Supplier Sigma Aldrich Supplier No. 38570025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Mercapto-5-benzimidazolesulfonic acid sodium salt dihydrate

Specifications

CAS 207511-11-3
Melting Point >300°C (lit.)
Molecular Formula C7H5N2NaO3S2 -+ 2H2O
Linear Formula C7H5N2NaO3S2 -+ 2H2O
MDL Number MFCD00192260
Quantity 100 mg
Molecular Weight (g/mol) 288.28
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.