Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercapto-5-thiazolidone 98.0+%, TCI America™
SDP

Catalog No. M02631G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M02631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M02631G Supplier TCI America Supplier No. M02631G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6913-23-1
Molecular Formula C3H3NOS2
MDL Number MFCD00059745
Synonym 2-Thio-5-thiazolidone

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 133.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Mercapto-5-thiazolidone
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.