Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercapto-6-(trifluoromethyl)-4-pyrimidinol 98.0+%, TCI America™
SDP

Catalog No. M27395G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27395G 5 g
M27391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27395G Supplier TCI America Supplier No. M27395G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 368-54-7
Molecular Formula C5H3F3N2OS
Molecular Weight (g/mol) 196.147
MDL Number MFCD00112978
InChI Key YZAOETRYQWFEOY-UHFFFAOYSA-N
Synonym 4-hydroxy-6-trifluoromethyl pyrimidine-2-thiol, 2-mercapto-6-trifluoromethyl pyrimidin-4-ol, 2-sulfanyl-6-trifluoromethyl pyrimidin-4-ol, 2-sulfanyl-6-trifluoromethyl-4-pyrimidinol, 4-hydroxy-2-mercapto-6-trifluoromethyl pyrimidine, 6-trifluoromethyl-2-thiouracil, 2-thioxo-6-trifluoromethyl-2,3-dihydropyrimidin-4 1h-one, 4-hydroxy-2-mercapto-6-trifluoromethylpyrimidine, uracil, 2-thio-6-trifluoromethyl, 2-mercapto-6-trifluoromethyl-pyrimidin-4-ol
PubChem CID 1239796
IUPAC Name 2-sulfanylidene-6-(trifluoromethyl)-1H-pyrimidin-4-one
SMILES C1=C(NC(=S)NC1=O)C(F)(F)F

Specifications

Melting Point 247°C
Color White-Yellow
Quantity 5 g
Formula Weight 196.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Mercapto-6-(trifluoromethyl)-4-pyrimidinol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.