Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptobenzothiazole, 97%

Catalog No. p-7021667
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1408622 100 g
AAA1408636 500 g
AAA140860E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1408622 Supplier Thermo Scientific Chemicals Supplier No. A1408622
Only null left
Add to Cart

CAS: 149-30-4 | C7H5NS2 | 167.244 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2-Mercaptobenzothiazole is used for detection of the equivalence point in volumetry (complexometry) and for concentration determinations in colorimetry. It is also used for the photometric determination of gold, cadmium, copper, lead and palladium. Further, it is used as a metalworking fluid, antifreeze, grease, anticorrosive agent, and adhesive. It acts as an antibacterial and antifungal. It is also employed in the rubber industry as a vulcanization accelerator. It inhibits dopamine beta-hydroxylase and biocorrosion in cooling systems, especially in the paper industry. In addition to this, it is used as a wood preservative and rubber additive.

Solubility
Soluble in ethanol, diethyl ether, benzene, carbon tetrachloride, alkali and alkaline solutions. Slightly soluble in acetone, naphtha and glacial acetic acid. Insoluble in water and gasoline.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-30-4
Molecular Formula C7H5NS2
Molecular Weight (g/mol) 167.244
MDL Number MFCD00005781
InChI Key YXIWHUQXZSMYRE-UHFFFAOYSA-N
Synonym 2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax
PubChem CID 697993
ChEBI CHEBI:34292
IUPAC Name 3H-1,3-benzothiazole-2-thione
SMILES C1=CC=C2C(=C1)NC(=S)S2

Specifications

Melting Point 178°C to 182°C
Density 1.42
Flash Point 243°C (469°F)
Quantity 100 g
UN Number UN3077
Beilstein 119484
Sensitivity Air sensitive
Merck Index 14,5868
Solubility Information Soluble in ethanol,diethyl ether,benzene,carbon tetrachloride,alkali and alkaline solutions; Slightly soluble in acetone,naphtha and glacial acetic acid; Insoluble in water and gasoline.
Formula Weight 167.25
Percent Purity 97%
Chemical Name or Material 2-Mercaptobenzothiazole
Show More Show Less
Hazard Category H317
Hazard Statement GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 205-736-8
RTECSNumber DL6475000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.