Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl borate, 99%, AcroSeal™

Catalog No. AC346128000
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle

CAS: 121-43-7 | C3H9O3B | 103.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-43-7
Molecular Formula C3H9O3B
Molecular Weight (g/mol) 103.91
MDL Number MFCD00008346
InChI Key WRECIMRULFAWHA-UHFFFAOYSA-N
Synonym trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3
PubChem CID 8470
ChEBI CHEBI:38913
IUPAC Name trimethyl borate
SMILES B(OC)(OC)OC

Specifications

Melting Point -34.0°C
Density 0.9150g/mL
Boiling Point 68.0°C to 69.0°C
Flash Point −8°C
Assay Percent Range 99%
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.3562 to 1.3582
Linear Formula (CH3O)3B
Quantity 800 mL
Beilstein 01, 287
Fieser 01,673; 13,218
Specific Gravity 0.915
Merck Index 15, 9882
Solubility Information Solubility in water: reacts. Other solubilities: miscible with thf,diethyl ether,hexane,methanol,,isopropylamine,and other organic liquids
Viscosity 0.36 mPa.s (25°C)
Formula Weight 103.91
Percent Purity 99%
Chemical Name or Material Trimethyl borate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-468-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.