Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptobenzothiazole, Spectrum™ Chemical
SDP

Catalog No. 18621104 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
25 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-104 25 kg
18-621-101 25 g
18-621-102 100 g
18-621-103 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-621-104 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M217125KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synonym: 2-Benzothiazolethiol, Captax, Benzo[d]thiazole-2(3H)-Thione, MBT

Chemical Identifiers

CAS 149-30-4

Specifications

Boiling Point >260°C (>500°F)
Packaging Drum
Quantity 25 kg
Molecular Weight (g/mol) 167.24
Percent Purity 96.0%
Grade Ungraded
Physical Form Powder or Waxy Solid
Chemical Name or Material 2-Mercaptobenzothiazole
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.