Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ 2-Mercaptoethanol

Catalog No. 21985023
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-985-023 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-985-023 Supplier Gibco™ Supplier No. 21985023
Only null left
Add to Cart
Add to Cart

A potent reducing agent used in cell culture medium to prevent toxic levels of oxygen radicals

  • Mercaptoethanol is not stable in solution. so most protocols require daily supplementation
  • Concentration of 55mM in Dulbecco's Phosphate Buffered Saline (DPBS)
  • Manufactured at a cGMP compliant facility, located in Grand Island, New York, registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 2°C to 8°C
Shipping conditions: Room temperature
Shelf life: 36 months from date of manufacture
Cell Type Mammalian Cells
Concentration 55 mM
Form Liquid
Product Type BME
pH 6 to 8
Quantity 50 mL
Shipping Condition Room Temperature
I've purchased Gibco 2-Mercaptoethanol (Cat. No. 21985023). Should I add it to my cell culture medium right before every use or can I add it to the bulk medium in advance?

The 2-Mercaptoethanol must be added to the medium immediately before use because activity rapidly declines in diluted form. You can supplement enough medium needed for that day.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.