Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Mercaptoethanol

Catalog No. M6250500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
100 mL
250 mL
500 mL
1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M6250500ML 500 mL
M6250100ML 100 mL
M625010ML 10 mL
M62501L 1 L
M6250250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. M6250500ML Supplier Sigma Aldrich Supplier No. M6250500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-mercaptoethanol is a thiol compound, commonly used as a reducing agent in organic reactions.bme is suitable for reducing protein disulfide bonds prior to polyacrylamide gel electrophoresis and is usually included in a sample buffer for sds-page at a concentration of 5%
  • Cleaving intermolecular (between subunits) disulfide bonds allows the subunits of a protein to separate independently on sds-page
  • Cleaving intramolecular (within subunit) disulfide bonds allows the subunits to become completely denatured so that each peptide migrates according to its chain length with no influence due to secondary structure

Specifications

CAS 60-24-2
Density 1.114 g/mL (at 25°C (literature))
Boiling Point 157°C (lit.)
Molecular Formula C2H6OS
Refractive Index n20/D 1.500 (literature)
Linear Formula HSCH2CH2OH
MDL Number MFCD00004890
Quantity 500 mL
Synonym beta-Mercaptoethanol; 2-Hydroxyethylmercaptan; BME; Thioethylene glycol
Molecular Weight (g/mol) 78.13
Percent Purity ≥99.0%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.