Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ 2-Mercaptoethanol, Electrophoresis Grade

Catalog No. 04806443 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-806-443 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-806-443 Supplier MP Biomedicals™ Supplier No. 04806443
Add to Cart
Add to Cart

2-Mercaptoethanol, Electrophoresis Grade reduces disulfide linkages in solubilizing proteins for gel electrophoresis.

  • 2-Mercaptoethanol is used to reduce disulfide linkages in solubilizing proteins for gel electrophoresis (typically used in SDS-PAGE sample buffer at 5% concentration).
  • Also reduces excess oxidative polymerization of catalysts.
  • Cleaving intermolecular (between subunits) disulfide bonds allows the subunits of a protein to separate independently on SDS-PAGE.
  • Cleaving intramolecular (within subunit) disulfide bonds allows the subunits to become completely denatured so that each peptide migrates according to its chain length with no influence due to secondary structure.
  • In solution, 2-mercaptoethanol is readily oxidized in air to a disulfide, especially at alkaline pH. Because of this property, it is widely used to protect proteins, enzymes in particular, from becoming inactive.
  • An excess of 2-mercaptoethanol (generally used at 0.01 M) will maintain the protein thiol groups in their reduced state.

Solubility

  • Miscible with water, alcohol, ether and benzene.
  • Solutions containing even trace levels (nanomolar) of some metal salts (especially copper (II) or cobalt(II)) are unstable at pH >5 and progressively less stable at higher pH values.
  • Most buffers (especially phosphate) contain enough metal salts to lead to substantial loss of -SH within 1 to 2 days.
  • The addition of EDTA helps to stabilize solutions.
  • If diluted product needs to be stored, it is recommended to store at pH 6 to 7 with EDTA (0.05 mM) at 2 to 8°C for no more than 2 to 3 days; however, we highly recommend adding neat 2-mercaptoethanol as needed to the sample.
TRUSTED_SUSTAINABILITY

Specifications

pH Range 5.0 (0.1 M in water)
Molecular Weight (g/mol) 78.13 g/mol
Purity ≥98%
CAS 60-24-2
Form Liquid
Boiling Point 157°C
Density 1.114 g/mL at 25°C
Flash Point 68°C (154.4°F)
Vapor Density 2.69
Vapor Pressure 1 mm Hg at 20°C
Quantity 25 g
Synonym β-Mercaptoethanol, 2-Hydroxyethylmercaptan, BME, Thioethylene glycol, 2-Hydroxy-1-ethanethiol, Monothioethyleneglycol
Solubility Miscible with water, alcohol, ether and benzene.
Melting Point <-50°C
Show More Show Less

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.