Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Mercaptoethanol, Electrophoresis Grade, ≥98%

Click to view available options
Quantity:
250g
1000g
60-24-2
C2H6OS
78.129
MFCD00004890
DGVVWUTYPXICAM-UHFFFAOYSA-N
2-mercaptoethanol,mercaptoethanol,thioglycol,beta-mercaptoethanol,ethanol, 2-mercapto,2-thioethanol,2-hydroxy-1-ethanethiol,thioethylene glycol,2-hydroxyethanethiol,2-hydroxyethyl mercaptan
1567
CHEBI:41218
2-sulfanylethanol
C(CS)O
60-24-2
C2H6OS
78.129
MFCD00004890
DGVVWUTYPXICAM-UHFFFAOYSA-N
2-mercaptoethanol,mercaptoethanol,thioglycol,beta-mercaptoethanol,ethanol, 2-mercapto,2-thioethanol,2-hydroxy-1-ethanethiol,thioethylene glycol,2-hydroxyethanethiol,2-hydroxyethyl mercaptan
1567
CHEBI:41218
2-sulfanylethanol
C(CS)O
Applications
β-Mercaptoethanol can act as an enzyme reactivator in systems necessitating reduction for activation, and has been commonly used to reduce disulfide bonds in order to separate protein subunits for use in electrophoresis. This product can also be used to form o-Phthalaldehyde-beta-mercaptoethanol (OPAME), a fluorogenic reagent shown to detect amino acids based on its reaction with primary amines. More recently OPAME has been used to distinguish between ssDNA/dsDNA and even single base mismatches in the hybridization of DNA. It is widely used for retarding oxidation of biological compounds in solution.

Solubility
Fully miscible in water. Soluble in alcohol, ether, benzene, and other organic solvents.

Notes
Handle under dry protective gas. Store in cool condition. Incompatible with air, moisture, oxidizing agents.
Boiling Point 156°C to 158°C
Odor Unpleasant
Sensitivity Air Sensitive & Hygroscopic
Color Colorless
Density 1.114g/mL
Melting Point -100°C
Quantity 250g
Linear Formula HSCH2CH2OH
UN Number UN2966
Beilstein 773648
Merck Index 14,5869
Formula Weight 78.13
Physical Form Liquid
Percent Purity ≥98%
Grade Electrophoresis
Chemical Name or Material 2-Mercaptoethanol
Show More Show Less

GHS H Statement
H301-H310-H318-H227-H315-H317-H335
Toxic if swallowed.
Fatal in contact with skin.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P210-P301+P310-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: II; Proper Shipping Name: THIOGLYCOL

RTECSNumber : KL5600000

TSCA : TSCA