Learn More
Sigma Aldrich 2-Mercaptoethyl Ethyl Sulfide Silica
Catalog No. 74511150G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-511-150G 50 g
74-511-110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 74-511-150G Supplier Sigma Aldrich Supplier No. 74511150G
Add to Cart
Add to Cart
Danger
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
2-Mercaptoethyl ethyl sulfide Silica is a mercaptoalkyl functionalized silica with metal scavenging ability.Silica Supported Metal Scavenger:Fe(III), Ni(0), Cu(II), Ru(III), Rh(I), Rh(III), Pd(0), Pd(II), Ag(I), Cd(II), Sn(II), Os, Ir(I), Ir(III), Pt(II), Au(I), Au(III), Hg(II), Pb(II)Sold in collaboration with PhosphonicS Ltd. for research purposes only.
Specifications
Quantity 50 g
Synonym PhosphonicS(TM) SEM26