Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptonicotinic acid, 90+%, Thermo Scientific™

Catalog No. AAA1468322
Change view
Click to view available options
Quantity:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1468322 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1468322 Supplier Thermo Scientific Chemicals Supplier No. A1468322

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 38521-46-9
Molecular Formula C6H5NO2S
Quantity 100g
MDL Number MFCD00010102
Beilstein 119029
Sensitivity Air Sensitive
Synonym 2-mercaptonicotinic acid, 2-sulfanylpyridine-3-carboxylic acid, 2-mercaptopyridine-3-carboxylic acid, 2-mercapto-nicotinic acid, unii-eu7d859abz, 3-pyridinecarboxylic acid, 1,2-dihydro-2-thioxo, 2-sulfanylnicotinic acid, 1,2-dihydro-2-thioxopyridine-3-carboxylic acid, eu7d859abz, 2-mercapto-3-pyridinecarboxylic acid
InChI Key WYKHFQKONWMWQM-UHFFFAOYSA-N
SMILES C1=CNC(=S)C(=C1)C(=O)O
IUPAC Name 2-sulfanylidene-1H-pyridine-3-carboxylic acid
Molecular Weight (g/mol) 155.171
PubChem CID 673681
Formula Weight 155.18
Percent Purity ≥90%
Chemical Name or Material 2-Mercaptonicotinic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.