Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Mesitylmagnesium Bromide Solution

Catalog No. 332402100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-240-2100ML 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 33-240-2100ML Supplier Sigma Aldrich Supplier No. 332402100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Mesitylmagnesium Bromide Solution

Specifications

CAS 19529-00-1
Melting Point 181°C to 183°C
Molecular Formula C72H66P4Ru
Linear Formula [(C6H5)3P]4RuH2
MDL Number MFCD04126429
Quantity 500 mg
Synonym Tetrakis(triphenylphosphine)ruthenium dihydride
Molecular Weight (g/mol) 1156.26 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.