Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich [2-(Methacryloyloxy)ethyl]dimethyl-(3-sulfopropyl)ammonium hydroxide

Catalog No. 537284250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-728-4250G 250 g
53-728-450G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 53-728-4250G Supplier Sigma Aldrich Supplier No. 537284250G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

[2-(Methacryloyloxy)ethyl]dimethyl-(3-sulfopropyl)ammonium hydroxide

Specifications

CAS 3637-26-1
Melting Point 150°C to 155°C
Molecular Formula C11H21NO5S
Linear Formula H2C=C(CH3)CO2CH2CH2N(CH3)2(CH2)3 SO3
Quantity 250 g
MDL Number MFCD00040574
Synonym DMAPS; 2-(N-3-Sulfopropyl-N,N-dimethyl ammonium)ethyl methacrylate; N-(3-Sulfopropyl)-N-(methacryloxyethyl)-N,N-dimethylammonium betaine
Molecular Weight (g/mol) 279.35
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.