Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich [2-(Methacryloyloxy)ethyl]trimethylammonium chloride solution

Catalog No. 408107250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-810-7250ML 250 mL
40-810-71L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-810-7250ML Supplier Sigma Aldrich Supplier No. 408107250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

[2-(Methacryloyloxy)ethyl]trimethylammonium chloride solution

Specifications

CAS 5039-78-1
Density 1.105 g/mL (at 25°C)
Molecular Formula C9H18ClNO2
Refractive Index n20/D 1.469
Linear Formula H2C=C(CH3)CO2CH2CH2N(CH3)3 Cl
Quantity 250 mL
MDL Number MFCD00060097
Synonym 2-Trimethylammonioethyl methacrylate chloride; 2-Trimethylammoniumethyl methacrylate chloride; Ethanaminium, N ,N ,N -trimethyl-2-[(2-methyl-1-oxo-2-propenyl)oxy]-chloride
Molecular Weight (g/mol) 207.7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.