Learn More
Medchemexpress LLC 2-Methacryloyloxyeth 250mg
SDP
Catalog No. 5000204610
Encompass_Preferred
2-Methacryloyloxyethyl phosphorylcholine is a polymer monomer containing a phospholipid polar group 2-Methacryloyloxyethyl phosphorylcholine polymers modify the surface properties of materials and reduce protein-surface interactions reduce IL-8 production inhibit cell Staphylococcus aureus and Porphyromonas gingivalis adhesion 2-Methacryloyloxyethyl phosphorylcholine is used in the study of medical device-associated infections periodontitis and dental caries[1][2][3][4]
Catalog No. 50-002-04610 Supplier Medchemexpress LLC Supplier No. 250MGHYW110551
2-Methacryloyloxyethyl phosphorylcholine is a polymer monomer containing a phospholipid polar group 2-Methacryloyloxyethyl phosphorylcholine polymers modify the surface properties of materials and reduce protein-surface interactions reduce IL-8 production inhibit cell Staphylococcus aureus and Porphyromonas gingivalis adhesion 2-Methacryloyloxyethyl phosphorylcholine is used in the study of medical device-associated infections periodontitis and dental caries[1][2][3][4]