Learn More
Medchemexpress LLC 2-methoxy-1,2-diphenylethanone | 3524-62-7 | MFCD00008492 | 98.1% | 226.27 g/mol | C15H14O2 | 1 ML

Supplier: Medchemexpress LLC HY11857110MM 1ML
Benzoin methyl ether (NSC76550) is a photoinitiator provided as a 10 mM solution in DMSO for laboratory photochemistry applications. The item is supplied as a 1 mL vial with high stated purity and referenced analytical documentation.
- 10 mM solution in DMSO, ready for use
- Pack size: 1 mL vial
- High purity (98.1% as stated on COA)
- Useful as a photoinitiator in polymerization and photochemical reactions
- Molecular formula C15H14O2; molecular weight 226.27 g/mol
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.