Learn More
Medchemexpress LLC 2-methoxy-13-methyl-6,7,8,9,11,12,14,15-octahydrocyclopenta[a]phenanthrene-3-carboxamide | 864668-87-1 | MFCD64668871 | 98.4% | 311.42 g·mol⁻¹ | C20H25NO2 | 10 MG
SDP
Catalog No. 5000288594
Encompass_Preferred
ENMD-1198 is an orally active, microtubule-destabilizing research compound and 2-methoxyestradiol analogue with antiproliferative and antiangiogenic activity. It is used in preclinical studies to investigate tubulin disruption, cell cycle arrest, apoptosis, and inhibition of hypoxia-inducible factor signaling.
  • Microtubule-destabilizing mechanism of action suitable for antiproliferative studies
  • Applicable in vitro and in vivo for antiangiogenic and tumor growth inhibition research
  • Available as a solid powder with high reported purity for research use
  • Soluble in DMSO at high concentration; documented in vivo formulation protocols for dosing
  • Powder and solution storage conditions provided to preserve stability and activity
Catalog No. 50-002-88594 Supplier Medchemexpress LLC Supplier No. HY1619610MG
ENMD-1198 is an orally active, microtubule-destabilizing research compound and 2-methoxyestradiol analogue with antiproliferative and antiangiogenic activity. It is used in preclinical studies to investigate tubulin disruption, cell cycle arrest, apoptosis, and inhibition of hypoxia-inducible factor signaling.
  • Microtubule-destabilizing mechanism of action suitable for antiproliferative studies
  • Applicable in vitro and in vivo for antiangiogenic and tumor growth inhibition research
  • Available as a solid powder with high reported purity for research use
  • Soluble in DMSO at high concentration; documented in vivo formulation protocols for dosing
  • Powder and solution storage conditions provided to preserve stability and activity