Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 2-Methoxy-3-pyridinecarboxaldehyde, 98%, [71255-09-9], F13069 137.136 C7H7NO2

Catalog No. 501409222
Encompass_Preferred

Compound: 2-Methoxy-3-pyridinecarboxaldehyde

CAS#: 71255-09-9

MDL#: not listed

Background: A methoxylated pyridine carbaldehyde used as a building block in pharmaceutical research. The aldehyde group provides a reactive handle for reductive amination and other carbonyl chemistry common in medicinal chemistry synthesis.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-140-9222 Supplier Frontier Specialty Chemicals Supplier No. F130691G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 2-Methoxy-3-pyridinecarboxaldehyde

CAS#: 71255-09-9

MDL#: not listed

Background: A methoxylated pyridine carbaldehyde used as a building block in pharmaceutical research. The aldehyde group provides a reactive handle for reductive amination and other carbonyl chemistry common in medicinal chemistry synthesis.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.