Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Methoxy-4,4,5,5-Tetramethyl-1,3,2-Dioxaborolane

Catalog No. 67394325G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-394-325G 25 g
67-394-35G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 67-394-325G Supplier Sigma Aldrich Supplier No. 67394325G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Methoxy-4,4,5,5-Tetramethyl-1,3,2-Dioxaborolane

Specifications

CAS 1195-66-0
Density 0.9642 g/mL at 25°C
Molecular Formula C7H15BO3
Linear Formula C7H15BO3
Refractive Index n20/D 1.4096
MDL Number MFCD03093086
Quantity 25 g
Synonym Methoxyboronic acid pinacol ester
Molecular Weight (g/mol) 158 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.