Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-4-methyl-5-nitropyridine, 95%, Thermo Scientific™

Catalog No. AAH6125803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6125803 1 g
AAH6125806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6125803 Supplier Thermo Scientific Chemicals Supplier No. H6125803

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Methoxy-4-methyl-5-nitropyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6635-90-1
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.15
MDL Number MFCD03095075
InChI Key DJNQRLCFAHKFLZ-UHFFFAOYSA-N
Synonym 2-methoxy-5-nitro-4-picoline, 2-methoxy-5-nitro-4-icoline, 2-methoxy-4-methyl-5-nitro-pyridine, pyridine, 2-methoxy-4-methyl-5-nitro, 2-methoxy-5-nitro-4-methylpyridine, pubchem6607, acmc-20ans4, ksc358g6j, 2-methoxy-4-methyl-5-nitropyridine
PubChem CID 243169
IUPAC Name 2-methoxy-4-methyl-5-nitropyridine
SMILES COC1=CC(C)=C(C=N1)[N+]([O-])=O

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 168.15
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Methoxy-4-methyl-5-nitropyridine
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.