Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-4-nitroaniline, 99%

Catalog No. p-7027456
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2422014 25 g
AAB2422022 100 g
AAB2422036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2422014 Supplier Thermo Scientific Chemicals Supplier No. B2422014
Only null left
Add to Cart

CAS: 97-52-9 | C7H8N2O3 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-52-9
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.15
MDL Number MFCD00007363
InChI Key GVBHRNIWBGTNQA-UHFFFAOYSA-N
Synonym fast red b base, benzenamine, 2-methoxy-4-nitro, 4-nitro-o-anisidine, fast red base b, fast red b, red b base, diazo fast red b, 2-amino-5-nitroanisole, azoamine pink o, fast red 5na base
PubChem CID 7337
IUPAC Name 2-methoxy-4-nitroaniline
SMILES COC1=CC(=CC=C1N)[N+]([O-])=O

Specifications

Melting Point 138°C to 142°C
Density 1.211
Quantity 25 g
Beilstein 879619
Formula Weight 168.15
Percent Purity 99%
Chemical Name or Material 2-Methoxy-4-nitroaniline
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301-H311-H341-H332
Toxic if swallowed.
Toxic in contact with skin.
Suspected of causing genetic defects.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 202-588-6
RTECSNumber BZ7170000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.