Learn More
Cayman Chemical 2-Methoxy-4-vInylphenol 5g
SDP
Catalog No. 502911812
Encompass_Preferred
A phenol with diverse biological activities; is a gut microbiota-derived active metabolite of ferulic acid; inhibits α-glucosidase, α-amylase, and tyrosinase (Kis = 47.7, 189, and 40.3 UG/ml, respectively); induces cell cycle arrest at the S and G2/M phases and apoptosis in HT-29 cells at 1 mM
Catalog No. 50-291-1812 Supplier Cayman Chemical Supplier No. 364275 G
Add to Cart
Add to Cart
A phenol with diverse biological activities; is a gut microbiota-derived active metabolite of ferulic acid; inhibits α-glucosidase, α-amylase, and tyrosinase (Kis = 47.7, 189, and 40.3 UG/ml, respectively); induces cell cycle arrest at the S and G2/M phases and apoptosis in HT-29 cells at 1 mM