Learn More
Medchemexpress LLC 2-methoxy-4-vinylphenol | 7786-61-0 | MFCD00015437 | 99.6% | 150.17 g/mol | C9H10O2 | 25 MG
SDP
Catalog No. 5000303928
Encompass_Preferred
2-Methoxy-4-vinylphenol is an analytical standard used for research and analytical applications. It is a methoxy-substituted vinylphenol (C9H10O2, MW 150.17 g/mol, CAS 7786-61-0) found in natural products and reported to have germination-inhibiting and anti-inflammatory properties. This high-purity reference material is intended for chromatographic calibration, method development, and compound identification in laboratory settings.
  • High purity suitable for analytical use.
  • Confirmed chemical identity and CAS registration.
  • Appropriate as a reference standard for chromatographic methods.
  • Relevant to flavor, fragrance, and biochemical research.
  • Provided as a research-use material with controlled handling recommendations.
Catalog No. 50-003-03928 Supplier Medchemexpress LLC Supplier No. 25MGHYW019940R
Add to Cart
Add to Cart
2-Methoxy-4-vinylphenol is an analytical standard used for research and analytical applications. It is a methoxy-substituted vinylphenol (C9H10O2, MW 150.17 g/mol, CAS 7786-61-0) found in natural products and reported to have germination-inhibiting and anti-inflammatory properties. This high-purity reference material is intended for chromatographic calibration, method development, and compound identification in laboratory settings.
  • High purity suitable for analytical use.
  • Confirmed chemical identity and CAS registration.
  • Appropriate as a reference standard for chromatographic methods.
  • Relevant to flavor, fragrance, and biochemical research.
  • Provided as a research-use material with controlled handling recommendations.