Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-methoxy-4-vinylphenol | 7786-61-0 | MFCD00015437 | 99.6% | 150.17 g/mol | C9H10O2 | 250 MG
SDP

Catalog No. 5000302865
Encompass_Preferred

2-methoxy-4-vinylphenol (CAS 7786-61-0) is an aromatic phenolic compound used as an analytical standard and research reagent, with applications in flavor and aroma studies and synthetic chemistry. It is supplied as a colorless to light-yellow liquid with high reported purity suitable for analytical use.

  • Chemical formula: C9H10O2.
  • Molecular weight: 150.17 g/mol.
  • Purity: 99.56% (as listed by manufacturer).
  • Physical state: colorless to light-yellow liquid; density ≈ 1.11 g/cm3.
  • Common applications include analytical standards, flavor and aroma research, and synthetic intermediates.
  • Available pack sizes: 1 g, 5 g, 25 g, 100 g, and larger quantities.

Catalog No. 50-003-02865 Supplier Medchemexpress LLC Supplier No. 250MGHYW019940R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-methoxy-4-vinylphenol (CAS 7786-61-0) is an aromatic phenolic compound used as an analytical standard and research reagent, with applications in flavor and aroma studies and synthetic chemistry. It is supplied as a colorless to light-yellow liquid with high reported purity suitable for analytical use.

  • Chemical formula: C9H10O2.
  • Molecular weight: 150.17 g/mol.
  • Purity: 99.56% (as listed by manufacturer).
  • Physical state: colorless to light-yellow liquid; density ≈ 1.11 g/cm3.
  • Common applications include analytical standards, flavor and aroma research, and synthetic intermediates.
  • Available pack sizes: 1 g, 5 g, 25 g, 100 g, and larger quantities.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.